Anti-Histone H3 Ser29 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
BOSSBS-3187R-A680
: Bioss
- Antibody type:Primary
- Antigen name:Histone H3 Ser29
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Form:Liquid
- Gene ID:8290
- Antigen synonyms:H3.4|HIST3H3|H3/t|H3FT|H3t|Histone H3.1t|H3/g
- Modification:Phosphorylation (Ser28)
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:15 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic phosphopeptide derived from human Histone H3 around the phosphorylation site of Ser29
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Core component of nucleosome. Nucleosomes wrap and compact DNA into chromatin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Histones thereby play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. DNA accessibility is regulated via a complex set of post-translational modifications of histones, also called histone code, and nucleosome remodeling.
Type: Primary
Antigen: Histone H3 Ser29
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Rat