Anti-MCT4 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-2698R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:MCT4
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:401-465/465
- Form:Liquid
- Gene ID:9123
- Antigen synonyms:Solute carrier family 16 member 3|MCT4|MCT-3|MCT 4|MCT-4|SLC16A3|MCT 3|MCT3|Monocarboxylate transporter 4
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:52 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human MCT4
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Proton-linked monocarboxylate transporter. catalyses the rapid transport across the plasma membrane of many monocarboxylates such as lactate, pyruvate, branched-chain oxo acids derived from leucine, valine and isoleucine, and the ketone bodies acetoacetate, beta-hydroxybutyrate and acetate (By similarity).
Type: Primary
Antigen: MCT4
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 401-465/465
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat