AFP Mouse Monoclonal Antibody [clone: A4] (ALEXA FLUOR® 680)
BOSSBSM-1622M-A680
: Bioss
- Antibody type:Primary
- Antigen name:Alpha-fetoprotein
- Antigen symbol:AFP
- Clonality:Monoclonal
- Clone:A4
- Conjugation:Alexa Fluor® 680
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:174
- Antigen synonyms:Alpha-fetoprotein|FETA|Alpha-fetoglobulin|AFPD|Alpha-1-fetoprotein|HPAFP|AFP
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:69 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:Full-length AFP protein
- Purification:Purified by Protein G
- Pack type:Vial
- Pk:100 µl
Binds copper, nickel, and fatty acids as well as, and bilirubin less well than, serum albumin. Only a small percentage (less than 2%) of the human AFP shows estrogen-binding properties.
Type: Primary
Antigen: Alpha-fetoprotein
Clonality: Monoclonal
Clone: A4
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range:
Host: Mouse
Isotype: IgG
Reactivity: Human