BrdU Mouse Monoclonal Antibody [clone: A7] (ALEXA FLUOR® 750)
BOSSBSM-0917M-A750
: Bioss
- Antibody type:Primary
- Antigen name:Bromodeoxyuridine
- Antigen symbol:BrdU
- Clonality:Monoclonal
- Clone:A7
- Conjugation:Alexa Fluor® 750
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:5-Bromo-2_-deoxyuridine|proliferation Marker|Bromodeoxyuridine|5-BrdU
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:BrdU conjugated to KLH
- Tested applications:ICC
- Purification:Purified by Protein G
- Pack type:Vial
- Pk:100 µl
Bromodeoxyuridine is a synthetic nucleoside that is an analogue of thymidine. BrdU is incorporated into DNA during S-phase of a cell cycle. It is commonly used in the detection of proliferating cells in living tissues
Type: Primary
Antigen: Bromodeoxyuridine
Clonality: Monoclonal
Clone: A7
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range:
Host: Mouse
Isotype: IgG
Reactivity: Human; Mouse; Rat; Others