Anti-PSMG1 Rabbit Polyclonal Antibody (ALEXA FLUOR® 750)
Catalog # BOSSBS-19577R-A750
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:PSMG1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:21-100/288
- Form:Liquid
- Gene ID:8624
- Antigen synonyms:PAC-1|C21-LRP|Down syndrome critical region protein 2|PAC1|C21LRP|LRPC21|Chromosome 21 leucine-rich protein|DSCR2|PSMG1|Proteasome assembly chaperone 1
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:33 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human PSMG1
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Chaperone protein which promotes assembly of the 20S proteasome as part of a heterodimer with PSMG2. The PSMG1-PSMG2 heterodimer binds to the PSMA5 and PSMA7 proteasome subunits, promotes assembly of the proteasome alpha subunits into the heteroheptameric alpha ring and prevents alpha ring dimerisation.
Type: Primary
Antigen: PSMG1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 21-100/288
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse