Anti-IFNA7 Rabbit Polyclonal Antibody (ALEXA FLUOR® 555)
BOSSBS-15521R-A555
: Bioss
- Antibody type:Primary
- Antigen name:IFNA7
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 555
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Epitope:121-189/189
- Form:Liquid
- Gene ID:3444
- Antigen synonyms:IFN alpha J1|IFNA7|OTTMUSP00000012102|IFNA 7|IFN-alpha-J1|IFNA7_HUMAN|Interferon alpha J|Interferon alpha J1|IFN alpha 7|IFN-alpha-7|Interferon alpha 7|Interferon alpha-J|RP23-139P14,26|LeIF J|IFNA J|IFN alphaJ|OTTHUMP00000021136|Interferon alpha family, gene 7|Interferon alpha-7|Interferon alpha-J1
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:20 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human IFNA7
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Produced by macrophages, IFN-alpha have antiviral activities. Interferon stimulates the production of two enzymes: a protein kinase and an oligoadenylate synthetase.
Type: Primary
Antigen: IFNA7
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 555
Public Immunogen Range: 121-189/189
Host: Rabbit
Isotype: IgG
Reactivity: