Anti-GPCR RTA Rabbit Polyclonal Antibody (ALEXA FLUOR® 680)
Catalog # BOSSBS-15372R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:GPCR RTA
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Epitope:51-150/343
- Form:Liquid
- Antigen synonyms:GPR168|mrgF|RTA|MRGPRF|MGC21621|GPR140|MRGRF_HUMAN
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:38 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human GPCR RTA
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
GPCR RTA.
Type: Primary
Antigen: GPCR RTA
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 51-150/343
Host: Rabbit
Isotype: IgG
Reactivity: