Order Entry
United Kingdom
ContactUsLinkComponent
Anti-DNA Polymerase Kappa/POLK Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anti-DNA Polymerase Kappa/POLK Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-13019R-A680
Supplier:  Bioss
Anti-DNA Polymerase Kappa/POLK Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-13019R-A680
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    DNA Polymerase Kappa/POLK
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 680
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    1-100/870
  • Form:
    Liquid
  • Gene ID:
    51426
  • Antigen synonyms:
    DNA polymerase kappa|POLK_HUMAN|Short=DINP|DINB protein
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
  • Molecular weight:
    99 kDa
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human DNA Polymerase Kappa/POLK
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

DNA polymerase lambda (pol Kappa), also designated DNA polymerase Kappa or Pol Beta2, is a low-fidelity polymerase which plays a role in both spontaneous and DNA damage-induced mutagenesis. Encoded by the POLL gene, pol Kappa is a member of the DNA polymerase type-X family. Pol Kappa extends primer-terminal mispairs opposite nondamaged DNA templates, suggesting that it may assist in extending mismatched base pairs during normal DNA replication. In addition, pol Kappa may play a role in the mutagenic bypass of T-T dimers. Proliferating cell nuclear antigen (PCNA), a protein essential to DNA replication, interacts with pol Kappa and thus influences the ability of pol Kappa to synthesize DNA.

Type: Primary
Antigen: DNA Polymerase Kappa/POLK
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 1-100/870
Host: Rabbit
Isotype: IgG
Reactivity: Human