Ampicillin sodium salt 91.0-102.0% (by anhydrous basis)
Catalog # MSPPA40040-25.0
Supplier: MARKET SOURCE PART PROCESS
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White Crystalline Powder
- Assay (Anhydrous Substance) :91.0 - 102 %
- Bacterial Endotoxins :≤ 0.15 IU/mg
- Dimethylaniline:≤ 0.002 %
- Identification:Conform
- Methylene Chloride :≤ 0.2 %
- pH (1% Solution):8.0 - 10.0
- Solubilty (50mg/ml in Water):Soluble
- Specific Optical Rotation:+258 - +287 °
- Sterility:Conform
- Water:≤ 2.0 %
Specification Test Results
About this item
Synonyms: D(-)-α-Aminobenzylpenicillin Sodium Salt, Ampicillin Na, Ampicillin sodium salt, D-(−)-α-Aminobenzylpenicillin sodium salt Ampicillin sodium salt
:
:371.39 g/mol
:GHS08
Storage Temperature:Refrigerator
CAS Number:69-52-3
Formula:C₁₆H₁₈N₃NaO₄S
MDL Number:MFCD00064313