Order Entry
United Kingdom
ContactUsLinkComponent
M9 Minimal Salts (Maniatis Formulation), Powder, MP Biomedicals
M9 Minimal Salts (Maniatis Formulation), Powder, MP Biomedicals
Supplier:  MP Biomedicals
Certificates
undefined
M9 Minimal Salts (Maniatis Formulation), Powder, MP Biomedicals
Supplier:  MP Biomedicals

Specifications

  • Sterility:
    A 56,4 g sample of M9 Minimal Salts was dissolved in water to 1 litre, the pH adjusted to 7,4, if needed, and the solution autoclaved to make a sterile 5X concentrate.
  • Application:
    Bacteria Cell Culture
  • pH:
    Acceptable pH range is: 6,8 ±0,2. (Lit.)

Specifications

Frequently Bought Together