Specifications
- Pk:96 Tests
- Assay Type:Sandwich
- Host:
- Primary antibody reactivity:
- Description:QuickDetect™ Catecholamine (CA) (Rat) ELISA Kit, 100 assays
- Environmentally Preferable:
Specifications
About this item
A rapid ELISA kit to quantify Catecholamine in Rat serum, plasma, urine, cell culture samples.
- Sensitivity: <1,2 pg/ml
- Cross Reactivity: No significant cross-reactivity or interference between this analyte and its analogues was observed
- Easy, convenient and time-saving method to measure the level of Catecholamine in rat serum, plasma, urine, cell culture samples, biological fluid
- Detection Range: 5 to 320 pg/ml
Catecholamine (CA) is a monoamine, an organic compound that has a catechol and a side-chain amine. Catecholamines are derived from the amino acid tyrosine, which is derived from dietary sources as well as synthesis from phenylalanine. Two catecholamines, norepinephrine and dopamine, act as neuromodulators in the central nervous system and as hormones in the blood circulation. The catecholamine norepinephrine is a neuromodulator of the peripheral sympathetic nervous system but is also present in the blood. It causes general physiological changes that prepare the body for physical activity. Some typical effects are increases in heart rate, blood pressure, blood glucose levels, and a general reaction of the sympathetic nervous system. BioVision’s Catecholamine ELISA kit is a sandwich ELISA assay for the quantitative measurement of CA in rat serum, plasma and cell culture supernatants in 90 minutes. The density of colour is proportional to the amount of Catecholamine captured from the samples.
All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.