- Pk:96 Tests
- Assay Type:Sandwich
- Host:
- Primary antibody reactivity:
- Description:QuickDetect™ GABA (Rat) ELISA Kit, 100 assays
- Environmentally Preferable:
A rapid ELISA kit for quantify of GABA in rat serum, plasma, urine, cell culture samples.
- Easy, convenient and time-saving method to measure the level of GABA in rat Serum, plasma, urine, cell culture samples, biological fluid
- Detection Range: 6 to 400 pg/ml
- Sensitivity: <1,2 pg/ml
- Cross Reactivity: No significant cross-reactivity or interference between this analyte and its analogues was observed
Gamma-Aminobutyric acid (γ-Aminobutyric acid) (GABA) is the chief inhibitory neurotransmitter in the mammalian central nervous system. Its principal role is reducing neuronal excitability throughout the nervous system. In vertebrates, GABA acts at inhibitory synapses in the brain by binding to specific transmembrane receptors in the plasma membrane of both pre- and postsynaptic neuronal processes. In rats, GABA is also directly responsible for the regulation of muscle tone. BioVision’s GABA ELISA kit is a sandwich ELISA assay for the quantitative measurement of GABA in rat serum, plasma and cell culture supernatants in 90 minutes. The density of colour is proportional to the amount of GABA captured from the samples.
All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.