Order Entry
United Kingdom
ContactUsLinkComponent
Bovine/Calf Blood in Heparin Sodium
Bovine/Calf Blood in Heparin Sodium
Catalog # ROCKR500-0100
Bovine/Calf Blood in Heparin Sodium
Catalog # ROCKR500-0100
Specifications
  • Description:
    Bovine/calf blood in heparin sodium
  • Shipping temperature:
    Wet ice
  • Pk:
    100 mL
Specifications
About this item
Bovine/calf blood in heparin sodium.
  • Aseptically collected and prepared to order
  • Blood draw in anticoagulant
Bovine/calf whole blood is aseptically collected and prepared to order from our colony of animals. These cell suspensions are useful for the titration of complement, adsorption procedures, testing for agglutinins/HA assays, and for the preparation of stroma as particulate reagents. Bovine/calf red blood cells are perishable and are collected and processed upon receipt of your order.
Washed pooled cells preparations, complement titration, adsorption procedures, HA assays and for the preparation of stroma as particulate reagents.