Sample pans, disposable
Disposable aluminium sample pans, PMB moisture balances
611-2916
Aluminum sample pans are ideal for moisture analysis testing.
The disposable pans fit neatly on the PMB moisture analysers pan holder. They are 10 mm deep to keep the items being weighed in place.
: This pack holds 250 disposable pans.
- For:PMB moisture balances
- Description:Disposable aluminium sample pans
- Seq:28