5,5'-(Hexafluoropropane-2,2-diyl)bis(2-(allyloxy)aniline) ≥95%
:2573217-07-7
:
:C₂₁H₂₀F₆N₂O₂
:MFCD32708902
:446.3861 g/mol
Frequently Bought Together
tert-Butyl methyl ether ≥99.5%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Benzene ≥99.5% (by GC), AnalaR NORMAPUR® ACS analytical reagent
: VWR Chemicals
o-Xylene ≥99.0% (by GC), Puriss. p.a., Riedel-de Haën™
: Honeywell Chemicals
Diethanolamine ACS
: Thermo Fisher Scientific
2-Propanol ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
: VWR Chemicals
Petroleum spirit, 30…40 °C, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
n-Pentane ≥99.0%, CHROMASOLV™ for HPLC, Riedel-de Haën™
: Honeywell Chemicals
n-Pentane, extra dry 99+%, AcroSeal®
: Thermo Fisher Scientific
4,4'-Bis(dimethylamino)diphenylmethane 98%
: Thermo Fisher Scientific
Methanol 99.9% for analysis
: Thermo Fisher Scientific
1-Chlorobutane 99.8% for HPLC
: Thermo Fisher Scientific
Hexane (mixture of isomers) ≥95%, Laboratory Reagent
: Honeywell Chemicals