Hydrogen chloride 5 - 10% (w/w) in methanol for esterification
FLUOX0041.500ML
:
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
- Appearance:Colorless to Almost colorless clear liquid
- Concentration (Neutralization titration):5.0 to 10.0 w/w%
:
:36.46 g/mol
:14,04780
:Refrigerator
:231-595-7
:2924
:3,II
:GHS02
:GHS05
:GHS07
:GHS08
:7647-01-0
:HCl
:MFCD00011324