Order Entry
United Kingdom
ContactUsLinkComponent
Furan-3-boronic acid (contains varying amounts of Anhydride)
Irritant
Furan-3-boronic acid (contains varying amounts of Anhydride)
Catalog # FLUOF0438.1G
Supplier:  FLUOROCHEM
CAS Number:  
Irritant
Furan-3-boronic acid (contains varying amounts of Anhydride)
Catalog # FLUOF0438.1G
Supplier:  FLUOROCHEM
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (Neutralization titration):
    97.0 to 119.0 %

Specification Test Results

About this item

 :
 :111.89 g/mol
 :GHS07
CAS Number:55552-70-0
Formula:C₄H₅BO₃
MDL Number:MFCD01319007