
4,5-Dimethyl-o-phenylenediamine ≥98.0% (by HPLC)
Catalog # FLUOD1083.25G
Supplier: FLUOROCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Gray to Brown powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 95.0 %
- Melting point:125.0 to 128.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
About this item
:
:136.2 g/mol
EINECS:221-635-1
:GHS07
CAS Number:3171-45-7
Formula:C₈H₁₂N₂
MDL Number:MFCD00007729