Order Entry
United Kingdom
ContactUsLinkComponent
4-(Methylsulphonyl)aniline hydrochloride ≥99%
Toxic
Irritant
4-(Methylsulphonyl)aniline hydrochloride ≥99%
Catalog # ABCRAB134063.0025
Supplier:  abcr
CAS Number:  
Toxic
Irritant
4-(Methylsulphonyl)aniline hydrochloride ≥99%
Catalog # ABCRAB134063.0025
Supplier:  abcr
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

About this item

CAS Number:177662-76-9
 :
Formula:C₇H₁₀ClNO₂S
MDL Number:MFCD00216490
 :207.67 g/mol
Melting Pt:237…239 °C
 :GHS06
 :GHS07
Storage Temperature:Ambient