Order Entry
United Kingdom
ContactUsLinkComponent
 
 
Chromium(III) picolinate ≥96.0% (by titrimetric analysis)
  ABCRAB141096.0025
 :  abcr
 :  
 
Chromium(III) picolinate ≥96.0% (by titrimetric analysis)
  ABCRAB141096.0025
 :  abcr
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 g

 

 

  • Appearance:
    Purple to Dark purple to Dark red powder to crystal
  • Purity (Iodometric Titration):
    min. 96.0 %

 

 

 :
 :14,02236
 :GHS07
 :14639-25-9
 :C₁₈H₁₂CrN₃O₆
 :MFCD00068715