Order Entry
United Kingdom
ContactUsLinkComponent
5-(Carboxymethyl)-5H-[1,2,4]triazino[5,6-b]indole-3-thiol
Irritant
5-(Carboxymethyl)-5H-[1,2,4]triazino[5,6-b]indole-3-thiol
Catalog # APOSOR13468-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
5-(Carboxymethyl)-5H-[1,2,4]triazino[5,6-b]indole-3-thiol
Catalog # APOSOR13468-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :260.27 g/mol
 :GHS07
CAS Number:309283-89-4
Formula:C₁₁H₈N₄O₂S
MDL Number:MFCD01820802