Deuterium oxide, powder
Catalog # SIAL151882-10X1ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 SET
- Seq:0125
Specifications
Specification Test Results
- Assay:≥98% (HPLC)
Specification Test Results
About this item
:
:20 g/mol
CAS Number:7789-20-0
Formula:D₂O
MDL Number:MFCD00044636