Order Entry
United Kingdom
ContactUsLinkComponent
Cadmium ≥99.5%
Cadmium ≥99.5%
Catalog # SIAL265365-100G
Supplier:  Merck
CAS Number:  
Cadmium ≥99.5%
Catalog # SIAL265365-100G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Assay:
    99.5% trace metals basis

Specification Test Results

About this item

 :
 :112.41 g/mol
Boiling Pt:767 °C (1013 hPa)
Melting Pt:321 °C
Density:8.64 g/cm³ (20 °C)
EINECS:231-152-8
UN:2570
ADR:6.1,II
 :GHS02
 :GHS06
 :GHS08
 :GHS09
CAS Number:7440-43-9
Formula:Cd
MDL Number:MFCD00010914