Simulated intestinal fluid USP Test Solution (TS)
Catalog # RICCR7109000-500A
Supplier: RICCA CHEMICAL
CAS Number:
Specifications
- Pack type:Poly bottle
- Pk:500 mL
- Seq:2725
Specifications
Specification Test Results
- Appearance:Clear to hazy with stringy fibers, colorless, characteristic odor
- pH at 25 °C (traceable to NIST SRM 185 & 186):6.8 ± 0.1 pH at 25.0 °C
Specification Test Results
About this item
:
Boiling Pt:~ 100 °C
Melting Pt:~ 0 °C
Density:1.0000 g/cm³
:GHS08
Storage Temperature:Ambient