Order Entry
United Kingdom
ContactUsLinkComponent
Ultra clean matting, Nomad™ 4300
Catalog # MMMASM45X18
Supplier:  3M
undefined
Ultra clean matting, Nomad™ 4300
Catalog # MMMASM45X18
Supplier:  3M
Specifications
  • Colour:
    White
  • Description:
    Entry mat 40 sheets/pad, 6 pads/case
  • L×W:
    1,15 m × 600 mm
  • Pk:
    6
Specifications
About this item
3M™ Nomad™ 4300 ultra clean matting is a transparent, disposable, multi-layered contamination control mat.
  • High tack surface removes dust and small particles from shoe soles and trolley wheels
  • Peel off one layer to expose the next
  • Lay onto any clean surface with no edging or holding frame required
  • Designed to help protect clean areas where cleanliness and hygiene are critical factors
  • Anti-microbial additive - Stops the growth of micro-organisms
3M™ Nomad™ ultra clean matting 4300 is a disposable, dust collecting floor mat consisting of 40 sheets coated with a pressure sensitive adhesive. High tack surface removes dust and small particles from shoe soles and trolley wheels. When the top layer is contaminated, it is then simply peeled off to expose the next fresh adhesive layer ready for immediate use. The mat can be laid directly onto any clean floor surface and being low profile it does not need any edging or frame. It is designed to help protect clean areas where cleanliness and hygiene are critical factors.