Order Entry
United Kingdom
ContactUsLinkComponent
Fittings for 10-32 Ports and ¹/₆" stainless steel tubing, EXP®
Fittings for 10-32 Ports and ¹/₆" stainless steel tubing, EXP®
Catalog # OPTT15-20-04128
Fittings for 10-32 Ports and ¹/₆" stainless steel tubing, EXP®
Catalog # OPTT15-20-04128
Specifications
  • Description:
    EXP® hand-tight 5 cm coupler, SS and PEEK
  • Seq:
    01
  • Pk:
    1
Specifications
About this item
The EXP® Fitting System is the premier adjustable nut and ferrule compression fitting for extreme high-pressure connections between ¹/₁₆" tubing and any 10 to 32 port.
  • The patent-pending titanium hybrid ferrule integrates the resilience of titanium with the proven sealing ability of PEEK in a superb one-piece design
  • The hybrid ferrule provides a perfect seal with every connection, yet the swage can be released without tools to adjust to the different port depths of various hardware
  • This adjustability provides unlimited value through repeated zero-dead-volume connections with any 10 to 32 port
  • With the EXP® fitting system, you no longer have to clip off and replace swaged ferrules or wonder if you are achieving the best possible connection