Form-Zero Formalin neutralizing powder
AZERES86631
- Capacity:500 cc
- Description:Form-Zero powder
- Pk:12
Form-Zero Powder neutralizes up to one gallon of 10% neutral buffered formalin waste.
- Neutralizes buffered formalin waste
- Reduced dusting and risk to lab personnel
- Unique formula eliminates drain-clogging polymers
One bottle (500 cc) of Form-Zero neutralizes up to one gallon of 10% neutral buffered formalin waste, or 4% glutaraldehyde
Non-hazardous, neutralized waste can be safely disposed after just 20 minutes with no pH adjustment. Unique formula eliminates drain-clogging polymers.
Container inserts directly in gallon container opening, reducing dusting and risk to lab personnel.