Order Entry
United Kingdom
ContactUsLinkComponent
Aflatoxin M1-solution 0.5 μg/mL in acetonitrile, Analytical standard, reference material, Fluka™
Aflatoxin M1-solution 0.5 μg/mL in acetonitrile, Analytical standard, reference material, Fluka™
  HONCR1960-2ML
 :  
Aflatoxin M1-solution 0.5 μg/mL in acetonitrile, Analytical standard, reference material, Fluka™
  HONCR1960-2ML
 :  

 

  • Pack type:
    Glass Bottle
  • Pk:
    2 mL
  • Standards:
    Analytical standard
  • Description:
    Aflatoxin M1-solution 0.5 μg/mL in acetonitrile
  • Storage conditions:
    -20°C
  • MDL No.:
    MFCD00871812
  • UN No.:
    1648
  • Synonym(s):
    (3R,7R)-3-hydroxy-11-methoxy-6,8,19-trioxapentacyclo[10.7.0.02,9.03,7.013,17]nonadeca-1,4,9,11,13(17)-pentaene-16,18-dione
  • Formula:
    C₁₇H₁₂O₇
  • Concentration:
    0.5 μg/mL in acetonitrile
  • Description:
    Aflatoxin M1
  • Flash point:
    5 °C
  • Physical form:
    Liquid

 

 

Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials  and organic reference materials.

  • Produced according to ISO 9001
  • Suitable for a wide range of industries and applications
  • Detailed Certificate of Analysis included with every delivery