Ethyl 2-trans-4-cis-decadienoate Analytical standard, reference material, Fluka™
Catalog # HONCR1918-1G
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:1 g
- Standards:Analytical standard
- CAS No.:3025-30-7
- Description:Ethyl 2-trans-4-cis-decadienoate
- Environmentally Preferable:
- Storage conditions:RT
- MDL No.:MFCD00082225
- Synonym(s):Ethyl (2E,4Z)-2,4-decadienoate
- Melting point:-60.3 °C
- Formula:C₁₂H₂₀O₂
- Molecular weight:196.29 g/mol
- Description:Ethyl 2-trans-4-cis-decadienoate
- Flash point:> 100 °C
- Physical form:Liquid
Specifications
About this item
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery