Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Penicillin V, Reference Standards

Penicillin V, Reference Standards

Supplier:   USP
Penicillin V, Reference Standards
Supplier:  USP
Certificates

2 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1504489.
  • 1504503.
  • Description:
  • Penicillin V
  • Penicillin V Potassium
  • CAS No.:
  • 87-08-1
  • 132-98-9
  • Formula:
  • C₁₆H₁₈N₂O₅S
  • C₁₆H₁₇KN₂O₅S

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1504489.
    Penicillin V
    Penicillin V
    USP reference standard
    Vial
    200 mg
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - £338.00
    £338.00
    1 * 200 mg
       £338.00

    Specifications:
    More
    Specifications
    Cat. No.
    1504489.
    Description
    Penicillin V
    CAS No.
    87-08-1
    Formula
    C₁₆H₁₈N₂O₅S
  • 1504503.
    Penicillin V Potassium
    Penicillin V Potassium
    USP reference standard
    Vial
    200 mg
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - £362.00
    £362.00
    1 * 200 mg
       £362.00

    Specifications:
    More
    Specifications
    Cat. No.
    1504503.
    Description
    Penicillin V Potassium
    CAS No.
    132-98-9
    Formula
    C₁₆H₁₇KN₂O₅S
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.