Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Sitagliptin System Suitability Mixtures, Reference Standard
Sitagliptin System Suitability Mixtures, Reference Standard
Sitagliptin System Suitability Mixture, USP reference standard, Vial, 20 mg
Catalog # 1612914.
Supplier:   USP
Sitagliptin System Suitability Mixtures, Reference Standard
Catalog # 1612914.
Supplier:  USP
CAS Number:  
Sitagliptin System Suitability Mixtures, Reference Standard
Sitagliptin System Suitability Mixture, USP reference standard, Vial, 20 mg
CAS Number:  
Price
£1,290.00
Log in for availability
1 * 20 mg
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Sitagliptin Phosphate containing about 0.2% of S-enantiomer
Certifications: US Pharmacopea Reference Standard
Documents
Specifications
Specifications
  • Pack type:
    Vial
  • Pk:
    20 mg
  • Standards:
    USP reference standard
  • Description:
    Sitagliptin System Suitability Mixture