Order Entry
United Kingdom
ContactUsLinkComponent
Sulpho-EGS (Ethylene glycol bis(sulphosuccinimidyl succinate)), Sigma-Aldrich®
Sulpho-EGS (Ethylene glycol bis(sulphosuccinimidyl succinate)), Sigma-Aldrich®
Catalog # SIAL803219-50MG
Supplier:  Merck
CAS Number:  
Sulpho-EGS (Ethylene glycol bis(sulphosuccinimidyl succinate)), Sigma-Aldrich®
Catalog # SIAL803219-50MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Seq:
    2370
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Sulfo EGS
 :
 :660.45 g/mol
 :M-Clarity™ quality level = MQ100
 :GHS07
Storage Temperature:Freezer
CAS Number:67410-92-6
Formula:C₁₈H₁₈O₁₈N₂Na₂S₂
MDL Number:MFCD00054988