Order Entry
United Kingdom
ContactUsLinkComponent
 
DSP (Dithiobis(succinimidyl propionate)), Pierce™
 
undefined
DSP (Dithiobis(succinimidyl propionate)), Pierce™

 

  • Environmentally Preferable:

 

 

  • Product Line:
    Pierce™
  • Labeling Method:
    Chemical Labeling
  • Crosslinker Type:
    Homobifunctional
  • Reactive Moiety:
    NHS Ester
  • Spacer:
    Cleavable, Long
  • Form:
    Powder
  • Quantity:
    50 mg / 1 g
  • Type:
    Crosslinker
  • Solubility:
    DMF, DMSO

 

Frequently Bought Together