Order Entry
United Kingdom
ContactUsLinkComponent
 
Sulpho-NHS-LC-Biotin, EZ-Link™, Pierce™
  PIER21335
 :  
undefined
Sulpho-NHS-LC-Biotin, EZ-Link™, Pierce™
  PIER21335
 :  

 

  • Pk:
    100 mg
  • Seq:
    2440

 

 

  • Description:
    EZ-Link Sulfo-NHS-LC-Biotin
  • Product Type:
    Sulfo-NHS-LC-Biotin
  • Quantity:
    100 mg
  • Product Line:
    EZ-Link™
  • Cell Permeability:
    Cell-Impermeant
  • Chemical Reactivity:
    Amine
  • Reactive Moiety:
    Active Ester, Succinimidyl Ester, Sulfo-NHS Ester
  • Solubility:
    DMF (Dimethylformamide), DMSO (Dimethylsulfoxide), Water
  • Label or Dye:
    Biotin
  • Label Type:
    Biotin & Analogs
  • Spacer:
    Mid-Length

 

 

 :
 :556.59 g/mol
 :Freezer
 :127062-22-0
 :C₂₀H₂₉N₄NaO₉S₂
 :MFCD00078534