
Cinacalcet hydrochloride
Catalog # ACRO466010010
Supplier: Thermo Fisher Scientific
CAS Number:
Specification Test Results
- Appearance:White to beige powder
- Infrared spectrum:Conforms
- HPLC:≥ 98.0%
- Water:≤ 0.5%
- Specific optical rotation:-32.0° to -24.0° (20°C, 589nm) (c=0.5 in methanol)
- Solubility:(2mg/ml in DMSO) Clear