

Azelnidipine
Catalog # ACRO465060010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Light yellow to yellow powder
- Infrared spectrum:Conforms
- Melting point:120°C to 126°C
- HPLC:≥ 97.5%
- Loss on drying:≤ 0.5%
Specification Test Results
About this item
:
Melting Pt:193.3°C…195°C
:582.66 g/mol
:GHS05
:GHS07
Storage Temperature:Ambient
CAS Number:123524-52-7
Formula:C₃₃H₃₄N₄O₆
MDL Number:MFCD00865803