Human CD93 / C1QR1 Protein (His Tag)
SINO12589-H08H-50
:
- Pk:50 µG
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:His Tag
- Storage conditions:–20…–80 °C
- Endotoxin content:<1,0 EU per μg of the protein as determined by the LAL method
- Biological activity:Immobilised Human CD93 (His Tag)(Cat:12589-H08H) at 2 μg/ml (100 μl/well) can bind Human IGFBP7 (hFc Tag)(cat:13100-H02H). The EC50 is 8 - 60 ng/ml.
- Gene ID:Q9NPY3
- Reconstitution instructions:A hardcopy of COA with reconstitution instruction is sent along with the products. Please refer to it for detailed information.
- Carrier-free:Y
- Animal-free:Y
- Protein synonyms:C1qR,C1qR(P),C1QR1,C1qRP,CDw93,dJ737E23.1,ECSM3,MXRA4
- Protein/peptide name:CD93
- Purity:>90% as determined by SDS-PAGE
- Molecular weight:The secreted recombinant human CD93 comprises 570 amino acids and has a predicted molecular mass of 59,6 kDa. As a result of glycosylation, rh CD93 migrates as an approximately 100 kDa band in SDS-PAGE under reducing conditions.
- Sequence:Met1-Lys580
- Formulation:Powder
- Shipping temperature:Ambient
A DNA sequence encoding the human MAP2K6 isoform 1 (P52564-1) (Met 1-Asp 334, Ser 207/Asp, Thr 211/Asp) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.