Anti-IgG2 beta Rabbit Polyclonal Antibody
Catalog # NOVUNBP1-72795
Supplier: Novus Biologicals
Specifications
- Antibody type:Secondary
- Antigen name:Immunoglobulin G2 beta
- Antigen symbol:IgG2b
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Mouse
- Western blot:Yes
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2
- Storage temperature:Store at 4 °C short term. Aliquot and store at -20 °C long term. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Mouse IgG2b heavy chain
- Purification:Immunogen affinity purified
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Rabbit Polyclonal Rabbit anti-Mouse IgG2b Antibody. Tested Applications: Western Blot, ELISA, Immunohistochemistry. Tested Reactivity: Mouse.
Type: Secondary
Antigen: IgG2 beta
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse