Anti-endothelial lipase mouse monoclonal antibody [clone: 1D9]
Catalog # NOVUNBP2-02396
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Endothelial Lipase
- Antigen symbol:Endothelial Lipase
- Clonality:Monoclonal
- Clone:1D9
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Gene ID:9388
- Antigen synonyms:EC 3.1.1|endothelial|EC 3.1.1.3|Endothelial cell-derived lipase|EDLendothelial lipase|ELlipoprotein lipase H|lipase|PRO719
- Storage buffer:PBS (pH 7,3), 1,0% BSA and 50% Glycerol
- Storage temperature:Store at 4 °C short term. Aliquot and store at −20 °C long term. Avoid freeze-thaw cycles.
- Immunogen:Full length human recombinant protein of human LIPG (NP_006024) produced in HEK293T cell.
- Purification:Immunogen affinity purified
- Size:100 μl
- Pk:0,1 mL
Specifications
About this item
The endothelial lipase antibody (1D9) from Novus Biologicals is a mouse monoclonal antibody to endothelial lipase. This antibody reacts with human. The endothelial lipase antibody (1D9) has been validated for the following applications: Western blot, flow cytometry, immunohistochemistry, immunocytochemistry / immunofluorescence, immunoprecipitation, immunohistochemistry-paraffin.
Type: Primary
Antigen: Endothelial Lipase
Clonality: Monoclonal
Clone: 1D9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human