Order Entry
United Kingdom
ContactUsLinkComponent
Parker Hannifin Compression Union Tees, PFA
Parker Hannifin Fitting, PFA, Compression Union Tee, 1/8" OD
Catalog # MFLX31320-29
CE certificate
undefined
Parker Hannifin Fitting, PFA, Compression Union Tee, 1/8" OD
Catalog # MFLX31320-29
CE certificate
Specifications
  • Connection Type:
    Compression Union
  • Flow pattern:
    T
  • Description:
    Compression Union Tee, PFA, ⅛"; 1/EA
  • Environmentally Preferable:
  • Sterilisation:
    Autoclave or ethylene oxide (ETO)
  • Warranty:
    18 months
  • Pk:
    1
  • Compression size / tube ext.Ø (port 3):
    ⅛"
  • Fitting material:
    PFA
Specifications
About this item
Get a secure seal at higher pressures.
  • Constructed of durable PFA
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
  • Connection Type:
    Compression Union
  • Flow pattern:
    T
  • Description:
    Compression Union Tee, PFA, ⅛"; 1/EA
  • Environmentally Preferable:
  • Sterilisation:
    Autoclave or ethylene oxide (ETO)
  • Warranty:
    18 months
  • Pk:
    1
  • Compression size / tube ext.Ø (port 3):
    ⅛"
  • Fitting material:
    PFA