Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Halsey Micro needle holder

Halsey Micro needle holder

Clamping length of 12,5 mm, Lockable, Stainless steel, 125 mm, Serrated, tungsten carbide, Straight, 1,4 mm
Catalog # FSCI12500-12
Marketsource
Supplier:   FINE SCIENCE TOOLS
Halsey Micro needle holder
Catalog # FSCI12500-12
Supplier:  FINE SCIENCE TOOLS
Marketsource
Halsey Micro needle holder
Halsey Micro needle holder

Clamping length of 12,5 mm, Lockable, Stainless steel, 125 mm, Serrated, tungsten carbide, Straight, 1,4 mm

Price
£119.98
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Type:
    Lockable
  • Description:
    Clamping length of 12,5 mm
  • Length:
    125 mm
  • Material:
    Stainless steel
  • Tip style:
    Serrated, tungsten carbide
  • Tip width:
    1,4 mm
  • Tip shape:
    Straight
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.