Bonn Artery scissors, ball tip
FSCI14086-09
- Type:Sharp/ball tip, straight tips, 1.5 mm ball diameter
- Length:90 mm
- Environmentally Preferable:
- Pk:1
- Blade length:13 mm
These scissors are made of stainless steel.
These scissors are made of stainless steel.