Liquid detergent, concentrated, Contrad 70
DECNCON705E
- Description:Contrad 70
- Pack type:Plastic container
- Pk:5 L
- pH range:>13
Surface active cleaning agent supplied as a concentrated liquid which is ideal for 'manual' cleaning and ultrasonic cleaners.
- Phosphate-free, alkaline
- Free rinsing
For cleaning and/or decontaminating glassware, ceramics, most plastics, stainless steel and ferous metals.
Milky white non-viscous liquid with faint odour. For dilution with water.
: Satisfies the criteria in BS EN 1040 1977 for bacterial activity.
: Not suitable for use with surfaces of aluminium or zinc. Not suitable for use with Polycarbonate.