Order Entry
United Kingdom
ContactUsLinkComponent
 
Multiple affinity removal column - Hu-6 (human)
Catalog # HEWL5185-5984
Supplier:  Agilent
Multiple affinity removal column - Hu-6 (human)
Catalog # HEWL5185-5984
Supplier:  Agilent

Specifications

  • Ø int.:
    4,60 mm
  • Description:
    Multiple affinity removal column - Hu-6 (Human)
  • Length:
    50 mm
  • Seq:
    01
  • Pk:
    1

Specifications

About this item

The multiple affinity removal LC column - Human 6 is designed as a simple way to remove six interfering high-abundant proteins from human plasma samples. Removal of these abundant proteins improves the subsequent LC/MS and electrophoretic analysis of the serum sample by effectively expanding the dynamic range of the analysis.

  • Column eliminates approximately 85 to 90% of total protein (albumin, IgG, IgA, transferrin, haptoglobin, and antitrypsin)
  • Serum injection volume ranges between 15 to 80 µl per run for the 4,6 mm I-Ø
  • Minimise sample loss by utilising our rapid, single injection protocols and optimised buffers
  • Columns can last for over 200 injections
  • Process the most plasma/serum over the column lifetime, up to 16000 µl for the 4,6 mm I-Ø columns

Protocols and column are optimised for use with human serum. Column not recommended for use with mouse serum. Column is compatible with most LC or HPLC pumps or instruments.