Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Human recombinant GRO-g
Human recombinant GRO-g
Human recombinant GRO-g
APOSBITC2015-1MG
:
Apollo Scientific
:
Human recombinant GRO-g
APOSBITC2015-1MG
:
Apollo Scientific
:
Pk:
1 mg
Protein/peptide type:
Recombinant
Species:
Human
Protein synonyms:
GRO-gamma, CXCL3
Protein/peptide name:
GRO-g