
2-Bromo-4,6-difluorobenzenesulphonyl chloride 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:351003-42-4
:
Formula:C₆H₂BrClF₂O₂S
MDL Number:MFCD03094561
:291.5 g/mol
Boiling Pt:265…267 °C
Density:1.93 g/cm³ (25 °C)
Flash Pt:>110 °C
Storage Temperature:Ambient
UN:3265
ADR:8,II
:GHS05
Frequently Bought Together