
3-Bromo-4-methylbenzotrifluoride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:66417-30-9
:
Formula:C₈H₆BrF₃
MDL Number:MFCD01631584
:239.04 g/mol
Boiling Pt:88 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together