1-Methyl-3-(trifluoromethyl)-5-{[3-(trifluoromethyl)phenyl]sulphanyl}-1H-pyrazole-4-carbaldehyde 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:400751-40-8
:
Formula:C₁₃H₈F₆N₂OS
MDL Number:MFCD01936008
:354.28 g/mol
Melting Pt:70…72 °C
Storage Temperature:Ambient
Frequently Bought Together