
N-Cyclopropyl-N-(4-piperidinyl)-3-(trifluoromethyl)benzenesulphonamide 95+%
Catalog # APOSPC9239-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:348.39 g/mol
Melting Pt:>195 °C (dec.)
:GHS07
Storage Temperature:Ambient
CAS Number:387350-79-0
Formula:C₁₅H₁₉F₃N₂O₂S
MDL Number:MFCD01568000
Melting Pt:>195 °C (dec.)