
2-Chloro-5-(trifluoromethyl)pyridine-4-carboxylic acid 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:505084-58-2
:
Formula:C₇H₃ClF₃NO₂
MDL Number:MFCD01862019
:225.55 g/mol
Melting Pt:185…186 °C (dec.)
:GHS07
Storage Temperature:Ambient
Frequently Bought Together